Merck
  • Long-Term Provision of Environmental Resources Alters Behavior but not Physiology or Neuroanatomy of Male and Female BALB/c and C57BL/6 Mice.
Long-Term Provision of Environmental Resources Alters Behavior but not Physiology or Neuroanatomy of Male and Female BALB/c and C57BL/6 Mice.
Journal of the American Association for Laboratory Animal Science : JAALAS (2015-12-04)
Amy E Clipperton-Allen, Joelle C Ingrao, Laura Ruggiero, Lucas Batista, Jelena Ovari, Jutta Hammermueller, John N Armstrong, Dorothee Bienzle, Elena Choleris, Patricia V Turner
ABSTRACT
Few studies have evaluated the long-term effects of providing environmental resources to mice. This consideration is important given that mice are often maintained in vivaria for months. We evaluated the effects of providing simple cage resources (wood wool, cotton nesting material, a plastic tunnel, and oat cereal) compared with standard housing (solid-bottom cage with hardwood chips) to group-housed adult male and female C57BL/6 and BALB/c mice (n = 20/sex/strain/group) over 6 mo to determine whether these resources had a lasting effect on animal physiology, anatomy, and behavior. Body weights increased in all groups over time but were proportionately higher in male and female BALB/c mice housed in resource-supplemented environments. Throughout the study, adding environmental resources had no effect on hematology and lymphocyte subsets, fecal corticoid metabolite levels, response to LPS injection, or dendritic spine length or density. Strain- or sex×environmentspecific changes occurred in dark-light activity and thermal nociceptive responses. Dominant agonistic behaviors, abnormal conspecific sexual behaviors, and social nonagonistic behaviors demonstrated sex and strain×environment interactions such that fewer maladaptive social behaviors were noted in mice that were provided with environmental resources. This association was particularly evident in male mice of both strains in resource-supplemented environments. A small but significant increase in brain weight:body weight ratios occurred in mice in resource-supplemented environments. Under the conditions evaluated here, consistent use of simple environmental resources had a positive long-term effect on the behavioral wellbeing of male and female BALB/c and C57BL/6 mice yet minimally affected other aspects of murine physiology and neuroanatomy.
Sigma-Aldrich
Potassium dichromate, 99.98% trace metals basis
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Potassium chromate, ≥99.95% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Potassium dichromate, BioXtra, ≥99.5%
Sigma-Aldrich
Mercury(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Potassium dichromate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Mercury(II) chloride, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chromate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, BioUltra, ≥99.5% (AT)