Merck
A new dimeric neolignan from Magnolia grandiflora L. seeds.
Archives of pharmacal research (2014-09-07)
Hong-Mei Li, Su-Rong Zhao, Qiang Huo, Tao Ma, Hao Liu, Jae Kyoung Lee, Young-Soo Hong, Cheng-Zhu Wu
ABSTRACT
Bioassay-guided fractionation of the MeOH extract of Magnolia grandiflora seeds resulted in the isolation of a new dimeric neolignan, named bishonokiol A (1), as well as two known neolignans magnolol (2) and honokiol (3). The structures of the compounds were determined on the basis of data obtained using NMR and MS. Bishonokiol A (1) showed potent anti-proliferative activities in four human cancer cell lines, with IC50 values ranging from 5.1 to 7.5 µM. Additionally, bishonokiol A (1) induced apoptosis, as well as down-regulated the expression of the anti-apoptotic protein Bcl-2 and caspase-3 cleavage in HepG2 cell line.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG