Merck
  • Assessing the usefulness of prostaglandin E2 (Cervidil) for transcervical artificial insemination in ewes.
Assessing the usefulness of prostaglandin E2 (Cervidil) for transcervical artificial insemination in ewes.
Theriogenology (2015-09-10)
Pawel M Bartlewski, Ivanka B R Candappa
ABSTRACT
The underlying theme of this study involved the evaluation of the dilatory effects of prostaglandin E2 on the ovine cervix and thus the assessment of its potential applicability to transcervical artificial insemination (TCAI) in ewes. A novel method of prostaglandin E2 administration (controlled slow-release vaginal inserts) was examined, and the practical implications of this approach including cervical penetrability and posttreatment pregnancy rates were evaluated. The Guelph method of TCAI was performed during the seasonal anestrus (n = 40) and the breeding season (n = 40) on multiparous Rideau Arcott × Polled Dorset ewes, with or without the pretreatment with Cervidil (for a duration of 12 hours or 24 hours before TCAI). Cervical penetration rates averaged 82.5% (66 of 80), and they varied neither (P > 0.05) between the two seasons nor between Cervidil-treated ewes and their respective controls. Cervidil priming significantly reduced the total time required for TCAI during the breeding season in comparison with controls (54 vs. 98 seconds), especially after the 24-hour exposure (38 vs. 108 seconds). The time taken to traverse the uterine cervix was negatively correlated (P < 0.05) with the breed (percentage of Rideau Arcott genotype) and lifetime lamb production in seasonally anestrous ewes. Four out of 36 (11%) successfully penetrated ewes in the breeding season (three ewes allocated to the 12-hour control group and one ewe that had received Cervidil for 12 hours) became pregnant and carried the lambs to term. Vaginal mucus impedance at TCAI was significantly and positively correlated with the total time required to complete the procedure in cyclic ewes, and the negative correlation between vaginal mucus impedance and total time values at the time of controlled intravaginal drug release device removal approached to significance in anestrous ewes. The present results indicate a moderate benefit of using Cervidil for inducing cervical dilation before TCAI in ewes, mainly in the breeding season. The specific reason(s) for impaired fertility after the TCAI using frozen-thawed ram semen remains to be elucidated.
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%