Merck
  • Angiotensin II inhibits Na+/K+ATPase activity in pulmonary artery smooth muscle cells via glutathionylation and with the involvement of a 15.6 kDa inhibitor protein.
Angiotensin II inhibits Na+/K+ATPase activity in pulmonary artery smooth muscle cells via glutathionylation and with the involvement of a 15.6 kDa inhibitor protein.
Indian journal of biochemistry & biophysics (2015-06-30)
Sayed Modinur Rahaman, Kuntal Dey, Tapati Chakraborti, Sajal Chakraborti
ABSTRACT
The role of angiotensin II in regulating Na+/K(+)-ATPase activity has been investigated in bovine pulmonary artery smooth muscle cells (BPASMCs). Our study reveals that angiotensin II inhibits the Na+/K+ATPase activity via glutathionylation of the pump with the involvement of an increase in NADPH oxidase-derived O2*-. Additionally, angiotensin II treatment to the cells increases the inhibitory potency of the 15.6 kDa inhibitor towards the Na+/K+ATPase activity.
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Hydroxyapatite, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Sigma-Aldrich
Calcium phosphate tribasic, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis