Merck
  • Advances and challenges in PBPK modeling--Analysis of factors contributing to the oral absorption of atazanavir, a poorly soluble weak base.
Advances and challenges in PBPK modeling--Analysis of factors contributing to the oral absorption of atazanavir, a poorly soluble weak base.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-04-15)
Mark Berlin, Aaron Ruff, Filippos Kesisoglou, Wei Xu, Michael Hong Wang, Jennifer B Dressman
ABSTRACT
Many active pharmaceutical ingredients (APIs) exhibit a highly variable pharmacokinetic (PK) profile. This behavior may be attributable to pre-absorptive, absorptive and/or post-absorptive factors. Pre-absorptive factors are those related to dosage form disintegration, drug dissolution, supersaturation, precipitation and gastric emptying. Absorptive factors are involved with drug absorption and efflux mechanisms, while drug distribution and clearance are post-absorptive factors. This study aimed to investigate the relative influence of the aforementioned parameters on the pharmacokinetic profile of atazanavir, a poorly soluble weakly basic compound with highly variable pharmacokinetics. The pre-absorptive behavior of the drug was examined by applying biorelevant in vitro tests to reflect upper gastrointestinal behavior in the fasted and fed states. The in vitro results were implemented, along with permeability and post-absorptive data obtained from the literature, into physiologically based pharmacokinetic (PBPK) models. Sensitivity analysis of the resulting plasma profiles revealed that the pharmacokinetic profile of atazanavir is affected by an array of factors rather than one standout factor. According to the in silico model, pre-absorptive and absorptive factors had less impact on atazanavir bioavailability compared to post-absorptive parameters, although active drug efflux and extraction appear to account for the sub-proportional pharmacokinetic response to lower atazanavir doses in the fasted state. From the PBPK models it was concluded that further enhancement of the formulation would bring little improvement in the pharmacokinetic response to atazanavir. This approach may prove useful in assessing the potential benefits of formulation enhancement of other existing drug products on the market.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium oleate, ≥82% (fatty acids), powder
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ammonium dihydrogenphosphate, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium oleate, ≥95% (capillary GC)
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium phosphate monobasic, suitable for plant cell culture
Sigma-Aldrich
Sodium oleate, ≥99%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.