Merck
  • A Comparative Matched Analysis of Clinical Outcomes between Transradial versus Transfemoral Percutaneous Coronary Intervention.
A Comparative Matched Analysis of Clinical Outcomes between Transradial versus Transfemoral Percutaneous Coronary Intervention.
The Israel Medical Association journal : IMAJ (2015-08-04)
Gabriel Greenberg, Tamir Bental, Eli I Lev, Abid Assali, Hanna Vaknin-Assa, Ran Kornowski
ABSTRACT
Several trials support the transradial route of percutaneous coronary intervention (PCI) since it reduces access-site vascular complications and bleeding. To examine the effects of transradial interventions (TRI) on clinical outcomes in a 'real-world' cohort of patients undergoing PCI. We analyzed 4873 consecutive patients who underwent PCI at a tertiary center and identified 373 patients who underwent TRI. Patients (radial vs. femoral) were compared using a propensity score analysis to best match between groups. Outcome parameters included total mortality, myocardial infarction (MI), repeat target vessel revascularization (TVR) rates, length of hospitalization and AHt/Hb/creatinine values during hospitalization. These were evaluated at 6 months and 1 to 3 years after PCI. The rates of major adverse cardiovascular event (MACE) and its constituents were similar in the transradial vs. transfemoral groups at all time intervals: 6.7% vs. 5.5% at 6 months, 10.3% vs. 10% at 1 year, 15.7% vs. 15% at 2 years, 15.7% vs. 16% at 3 years, respectively (P = 0.6). The length of hospitalization was shorter in the TRI group (2.87 days ± 2.04 vs. 3.3 days ± 3.12, P = 0.023). We did not find significant differences between the groups in the mean AHt/Hb/creatinine values during the hospitalization course. In a real-world setting of PCI, the TRI route of PCI is as safe and efficient as the femoral approach. TRI is associated with shorter duration of hospitalization.
Sigma-Aldrich
Prasugrel, ≥98% (HPLC)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Bivalirudin trifluoroacetate salt, ≥97% (HPLC)
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%