Merck
  • L-Arginine Transport and Nitric Oxide Production in Kinin Receptor B1-/- Endothelial Cells.
L-Arginine Transport and Nitric Oxide Production in Kinin Receptor B1-/- Endothelial Cells.
Protein and peptide letters (2015-10-09)
Renato Cardoso Tudela, Rodrigo Azevedo Loiola, Tathiany Corteze Torres, Noemi Lourenço Gil, Nilson Antonio Assunção, Samuel M Ribeiro de Noronha, Silvana Aparecida Correa-Noronh, Richardt Gama Landgraf, Liliam Fernandes
ABSTRACT
Kinins are important vasoactive peptides, but the role of the B1 receptor subtype in the vascular control is poorly understood. This study analyzed the nitric oxide (NO) release, L-arginine (L-Arg) uptake and the expression of the cationic amino acid transporter (CAT) -1 in endothelial cells obtained from B1 receptor knockout (B1-/-) and wild type (WT) mice. NO production was assessed through a fluorescent dye in living cells stimulated with acetylcholine. L-Arg uptake was determined indirectly in the culture medium by HPLC, in the presence or absence of the CAT-1 blocker N-ethylmaleimide (NEM). CAT-1 mRNA levels and protein expression were determined by qPCR and western blot, respectively. NO release was significantly reduced in B1-/- when compared to WT cells. This result was accompanied by a decreased rate in the L-Arg uptake by B1-/- cells. Incubation with NEM impaired the L-Arg uptake in WT, but had no effect in B1-/- cells. Protein expression and mRNA levels for CAT-1 were reduced in B1-/- in comparison to WT cells. These findings suggest an important role of the endothelial B1 receptor in the vascular control by interfering with CAT-1 expression, L-Arg uptake and NO release.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Phthaldialdehyde, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O