Merck
  • A Multidisciplinary Assessment of River Surface Water Quality in Areas Heavily Influenced by Human Activities.
A Multidisciplinary Assessment of River Surface Water Quality in Areas Heavily Influenced by Human Activities.
Archives of environmental contamination and toxicology (2015-04-03)
Nikolina Charalampous, Aggeliki Kindou, Dimitris Vlastos, Vasiliki Tsarpali, Maria Antonopoulou, Ioannis Konstantinou, Stefanos Dailianis
ABSTRACT
The present study could serve as a multidisciplinary approach for the assessment of river surface water quality with the use of chemical and biological methods. Specifically, physicochemical parameters, heavy metals, and pesticides were measured in water samples from three different stations (sampling station S1, S2, and S3) along Asopos River (Greece). In parallel, algal species (primary producers)-such as Scenedesmus rubescens and Chlorococcum sp.; consumer invertebrate species, such as the fairy shrimp Thamnocephalus platyurus and the rotifer Brachionus calyciflorus; as well as human lymphocytes-were exposed to those samples for assessing their toxic and genotoxic/mutagenic effects. According to the results, although the values of almost all of the physicochemical parameters tested, heavy metals (zinc, cadmium, lead, and mercury) and pesticides were lower than or within the respective environmental quality standards, thus offering no clear evidence for their natural or anthropogenic origin. Values recorded for nickel, chromium, hexavalent chromium, and malathion represent a typical case of mixed influence from natural and anthropogenic enrichments. In contrast, the algal growth arrest, the acute toxic effects on the freshwater invertebrates, and the increased micronuclei frequencies observed in human lymphocytes showed the presence of human-derived hazardous substances, which were hardly determinable with the use of conventional chemical methods. Given that the presence of priority pollutants in river surface waters, heavily burdened by anthropogenic activities, could give no clear evidence for their biological risk, the results of the present study showed that chemical and biological assays should be applied in parallel, thus serving as a reliable tool for the assessment of river water quality.