Merck
  • Roles of DPY30 in the Proliferation and Motility of Gastric Cancer Cells.
Roles of DPY30 in the Proliferation and Motility of Gastric Cancer Cells.
PloS one (2015-07-07)
Yong Joo Lee, Myoung-Eun Han, Su-Jin Baek, Seon-Young Kim, Sae-Ock Oh
ABSTRACT
Various types of histone methylation have been associated with cancer progression. Depending on the methylation site in histone proteins, its effects on transcription are different. DPY30 is a common member of SET1/MLL histone H3K4 methyltransferase complexes. However, its expression and roles in gastric cancer have been poorly characterized. To determine whether DPY30 has pathophysiological roles in gastric cancer, its expression and roles were examined. Immunohistochemistry and real time PCR showed up-regulation of DPY30 expression in some gastric cancer cell lines and patients' tissues. Its knockdown by siRNA decreased the proliferation, migration, and invasion of gastric cancer cells, whereas its overexpression showed the opposite effects. These results indicate that DPY30 has critical roles in the proliferation, migration, and invasion of gastric cancer cells, and suggest DPY30 might be a therapeutic target in gastric cancer.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human DPY30
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%