Merck
  • EDTA functionalized magnetic nanoparticle sorbents for cadmium and lead contaminated water treatment.
EDTA functionalized magnetic nanoparticle sorbents for cadmium and lead contaminated water treatment.
Water research (2015-05-23)
Yuxiong Huang, Arturo A Keller
ABSTRACT
Cadmium (Cd(2+)) and lead (Pb(2+)) are toxic to human beings and other organisms, and the U.S. Environmental Protection Agency (EPA) has classified both as probable human carcinogens. In this study, a regenerable magnetic ligand particle (Mag-Ligand) which includes a metal-binding organic ligand (EDTA) attached to an iron oxide nanoparticle was developed for rapid removal of Cd(2+) and Pb(2+) as well as other metals from contaminated water. Mag-Ligand showed fast removal ability for both Cd(2+) (<2 h) and Pb(2+) (<15 min) with relatively high sorption capacity (79.4 and 100.2 mg/g for Cd(2+) and Pb(2+), respectively). The removal performance of Mag-Ligand was high across a wide pH range (3-10) as well as in the presence of competitive metal ions (Ca(2+) and Mg(2+)). In addition, Mag-Ligands can be easily regenerated (washed by 1% HCl) and reused several cycles with high sorption capacity. This study indicated that Mag-Ligand is a reusable sorbent for rapid, convenient, and efficient removal of Cd(2+) and Pb(2+) from contaminated aquatic systems.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)