Merck
  • Efficient Total Synthesis of Bongkrekic Acid and Apoptosis Inhibitory Activity of Its Analogues.
Efficient Total Synthesis of Bongkrekic Acid and Apoptosis Inhibitory Activity of Its Analogues.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-02)
Kenji Matsumoto, Masaki Suyama, Satoshi Fujita, Takuya Moriwaki, Yukiko Sato, Yoshifumi Aso, Satoshi Muroshita, Hiroshi Matsuo, Keishi Monda, Katsuhiro Okuda, Masato Abe, Hiroyuki Fukunaga, Arihiro Kano, Mitsuru Shindo
ABSTRACT
Bongkrekic acid (BKA), isolated from the bacterium Burkholderia cocovenenans, is an inhibitor of adenine nucleotide translocator, which inhibits apoptosis, and is thus an important tool for the mechanistic investigation of apoptosis. An efficient total synthesis of BKA has been achieved by employing a three-component convergent strategy based on Kocienski-Julia olefination and Suzuki-Miyaura coupling. It is noteworthy that segment B has been prepared as a new doubly functionalized coupling partner, which contributes to shortening of the number of steps. Torquoselective olefination with an ynolate has also been applied for the efficient construction of an unsaturated ester. Furthermore, it is revealed that 1-methyl-2-azaadamantane N-oxyl is an excellent reagent for final oxidation to afford BKA in high yield. Based on the total synthesis, several BKA analogues were prepared for structure-activity relationship studies, which indicated that the carboxylic acid moieties were essential for the apoptosis inhibitory activity of BKA. More easily available BKA analogues with potent apoptosis inhibitory activity were also developed.
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free