Merck
  • Medication beliefs are associated with phosphate binder non-adherence in hyperphosphatemic haemodialysis patients.
Medication beliefs are associated with phosphate binder non-adherence in hyperphosphatemic haemodialysis patients.
British journal of health psychology (2014-09-12)
Vari Wileman, Ken Farrington, David Wellsted, Mike Almond, Andrew Davenport, Joseph Chilcot
ABSTRACT
Patients with end-stage kidney disease receiving haemodialysis (HD) are at risk of cardiovascular disease and bone disorders related to high levels of serum phosphate. We studied the association between medication beliefs and depressive symptoms, with non-adherence to phosphate binding medication in a group of HD patients at risk of complications due to hyperphosphatemia. Cross-sectional design. Baseline data from 112 patients participating in a randomized controlled trial, evaluating an adherence intervention, are presented. All patients had serum phosphate levels >1.6 mmol/l at baseline. Adherence was measured by (1) serum phosphate and (2) Medication Adherence Report Scales (MARS). Beliefs about Medicines (BMQ) and depressive symptoms (PHQ-9) were also evaluated. Beliefs about Medicines Questionnaire necessity, but not concerns, beliefs were found to correlate with serum phosphate (r = -.23, p < .05) and self-reported adherence (r = .35, p < .01). In regression models, controlling for demographic, clinical and psychological variables, necessity beliefs explained the variance of serum phosphate (β = -.22, p = .01) and self-reported adherence (β = .30, p ≤ .01). Both BMQ concerns and depressive symptoms were not related to non-adherence. Patients' beliefs about the necessity of their prescribed phosphate binding medications explain variation in non-adherence levels, measured both subjective and objectively. Dialysis patient's medication beliefs are potentially modifiable targets for future interventions.
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis