Merck
  • Colloidal properties and behaviors of 3 nm primary particles of detonation nanodiamonds in aqueous media.
Colloidal properties and behaviors of 3 nm primary particles of detonation nanodiamonds in aqueous media.
Physical chemistry chemical physics : PCCP (2015-06-03)
N O Mchedlov-Petrossyan, N N Kamneva, A I Marynin, A P Kryshtal, E Ōsawa
ABSTRACT
This study was aimed to reveal the principal colloidal properties of the aqueous dispersion of extremely small primary single-crystalline diamond particles in water. Together with the non-diamond layer, the size of the colloidal species is 2.8 ± 0.6 nm as found via DLS of the initial 5.00 wt/vol% hydrosol. Anionic dyes are readily adsorbed on the colloidal species. This is in line with the positive zeta-potential. The critical coagulation concentrations of the 0.19 wt/vol% nanodiamond hydrosol were determined with a set of inorganic electrolytes and anionic surfactants. The data are in line with the Schulze-Hardy rule for "positive" sols. The fulfillment of the lyotropic (Hofmeister) series was also observed for single-charged anions. The abnormal influence of alkali gives evidence of the acidic nature of the positive charge of the nanodiamond species. Application of acid-base indicators allows estimating the value of the interfacial electrical potential of the nanodiamond particles. Upon dilution from 5.00% to 0.01%, the colloidal system under study exhibits unusual changes. The average size increases ca. ten-fold as determined by DLS. The TEM images support this observation. At the same time, the viscosity drops. This phenomenon was explained in terms of the so-called periodic colloidal structures (colloidal crystals) in concentrated solutions.
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Bromocresol Green Sultone Form
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Bromocresol Green, ACS reagent, Dye content 95 %
Sigma-Aldrich
Bromocresol Purple, BioReagent, suitable for indicator, Dye content 90 %
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
Bromocresol Purple, Technical grade
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)