Merck
  • MicroRNA dynamics during human embryonic stem cell differentiation to pancreatic endoderm.
MicroRNA dynamics during human embryonic stem cell differentiation to pancreatic endoderm.
Gene (2015-08-25)
Gary B Fogel, Zoya S Kai, Sahar Zargar, Andrew Hinton, G Adam Jones, Augusta S Wong, Sevan G Ficici, Ana D Lopez, Charles C King
ABSTRACT
MicroRNAs (miRNAs) are small non-coding RNAs that have emerged as critical regulators of human embryonic stem cell (hESC) pluripotency and differentiation. Despite the wealth of information about the role individual that miRNAs play in these two processes, there has yet to be a large-scale temporal analysis of the dynamics of miRNA expression as hESCs move from pluripotency into defined lineages. In this report, we used Next Generation Sequencing (NGS) to map temporal expression of miRNAs over ten 24-hour intervals as pluripotent cells were differentiated into pancreatic endoderm. Of the 2042 known human miRNAs, 694 had non-zero expression on all 11 days. Of these 694 miRNAs, 494 showed statistically significant changes in expression during differentiation. Clusters of miRNAs were identified, each displaying unique expression profiles distributed over multiple days. Selected miRNAs associated with pluripotency/differentiation (miR-302/367 and miR-371/372/373) and development/growth (miR-21, miR-25, miR-103, miR-9, and miR-92a) were found to have distinct expression profiles correlated with changes in media used to drive the differentiation process. Taken together, the clustering of miRNAs to identify expression dynamics that occur over longer periods of time (days vs. hours) provides unique insight into specific stages of differentiation. Major shifts in defined stages of hESC differentiation appear to be heavily dependent upon changes in external environmental factors, rather than intrinsic conditions in the cells.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder