Merck
  • Concentrations of luteinizing hormone and ovulatory responses in dairy cows before timed artificial insemination.
Concentrations of luteinizing hormone and ovulatory responses in dairy cows before timed artificial insemination.
Journal of dairy science (2015-07-06)
S L Pulley, D H Keisler, J S Stevenson
ABSTRACT
The objective was to determine the incidence of LH surges and ovulatory responses in lactating dairy cows enrolled in a timed artificial insemination (TAI) program. Cows were assigned randomly to 2 presynchronization treatments: (1) Pre10 (n=37): 2 injections of PGF2α (PG; PG-1 and PG-2) 14 d apart (Presynch); or (2) PG-3-G (n=33): one 25-mg injection of PG (Pre-PG) administered 3 d before a 100-µg GnRH injection (Pre-GnRH). Ten days after PG-2 or Pre-PG, all cows were enrolled in a 7-d Ovsynch TAI program [injection of GnRH (GnRH-1) 7 d before PG (PG-3) and GnRH (GnRH-2) administered at either 56 or 72 h after PG-3; TAI at 72 h]. Blood was collected to determine LH at (1) Pre-GnRH: 48 to 80 h after PG-2 and hourly from 72 to 78 h (Pre-GnRH at 72 h); (2) GnRH-1: 0 to 6 h after GnRH-1; and (3) GnRH-2: 48 to 80 h after PG-3 and hourly from 56 to 62 h or 72 to 78 h for cows injected with GnRH-2 at 56 or 72 h after PG-3, respectively. Ovaries were scanned and pregnancy per TAI (P/AI) was diagnosed 31 and 61 d post-TAI by transrectal ultrasonography. The Pre-GnRH injection increased the incidences of LH surges (100 vs. 43%) and ovulation (91 vs. 60%) and subsequent concentrations of progesterone in PG-3-G cows compared with Pre10 cows, respectively. Seven days later, incidence of ovulation (48 to 62%) and occurrence of LH surges (100%) did not differ between treatments after GnRH-1. In contrast, LH concentrations and area under the LH curve of Pre10 cows were greater than that of PG-3-G cows because progesterone was greater in PG-3-G than in Pre10 cows (4.6±0.4 vs. 2.8±0.4 ng/mL), respectively. Concentrations of LH did not differ after GnRH-2 at either 56 or 72 h; however, 1 cow receiving GnRH-2 at 56 h and 3 cows at 72 h had early spontaneous LH surges before GnRH-2. Ovulation was suppressed overall in 210 blood collection windows in cows with elevated progesterone concentrations. When progesterone was <1 ng/mL after either PG-2 or PG-3 injections, GnRH-induced LH surges occurred in more than 90% of cows, and incidence of ovulation exceeded 80%. Pregnancy per AI tended to differ for PG-3-G (56.7%) compared with Pre10 (37.8%) and for 56 h (54.5%) compared with 72 h (38.2%), with the Pre10-72 h treatment combination producing less than half (22.2%) the pregnancies compared with all other treatment combinations. Furthermore, in these same cows, post-TAI luteal tissue volume tended to be compromised. We conclude that incidences of GnRH-induced LH surges and ovulation are suppressed in cows with elevated progesterone, possibly contributing to some loss in P/AI in TAI programs.
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, meets USP testing specifications