Merck
  • Online 2D-LC-MS/MS assay to quantify therapeutic protein in human serum in the presence of pre-existing antidrug antibodies.
Online 2D-LC-MS/MS assay to quantify therapeutic protein in human serum in the presence of pre-existing antidrug antibodies.
Analytical chemistry (2015-08-04)
Yinghua Shen, Guodong Zhang, Jinsong Yang, Yongchang Qiu, Thomas McCauley, Luying Pan, Jiang Wu
ABSTRACT
The formation of antidrug antibodies (ADA) can interfere with the accurate quantitation of therapeutic proteins, leading to significantly underestimated drug concentrations and confounded pharmacokinetic (PK) data interpretation. Although highly desirable, development of ADA-tolerant bioanalytical methods enabling unbiased measurement of both free and ADA-bound drug presents a considerable challenge. We report herein the development and validation of a robust LC-MS assay capable of quantifying therapeutic protein immunoglobulin A1 protease (IgAP) in human serum in the presence of pre-existing anti-IgAP antibodies. The procedure included sodium dodecyl sulfate (SDS) denaturation and chemical reduction of serum proteins to dissociate ADA-drug bindings, followed by tryptic digestion of protein pellets and subsequent LC-MS analysis of the surrogate IgAP peptide using stable isotope labeled peptide internal standard. Substantial enhancements in the sensitivity and selectivity were achieved by the combination of online two-dimensional reversed-phase LC (2D-LC) operated in high and low pH buffers, respectively, for efficient enrichment and quantitation of the surrogate peptide by multiple-reaction monitoring (MRM) mass spectrometry. Unlike ligand-binding assay, our method is not prone to interferences from ADA, allowing accurate and precise measurement of the IgAP in the range of 0.05 to 10 μg/mL in 25 μL of human serum with a wide range of anti-IgAP antibody levels. The intra- and inter-run precision (coefficient of variation (CV%)) was within 11.5% and 10.5%, respectively, and the bias was within ±7.1% for all quality control (QC) concentrations. With little modification, the described method can readily be applicable to the quantitation of other biotherapeutic proteins in the ADA-positive clinical matrices.
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
SAFC
Iodoacetamide
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis