Merck
  • Chronic schizophrenia is associated with over-expression of the interleukin-2 receptor gamma gene.
Chronic schizophrenia is associated with over-expression of the interleukin-2 receptor gamma gene.
Psychiatry research (2014-04-10)
Hovsep Ghazaryan, Martin Petrek, Anna Boyajyan
ABSTRACT
Altered immune response, including low-grade inflammatory processes, is involved in the pathogenesis of schizophrenia, a chronic psychiatric disorder with complex etiology. Distinct gene variants of a number of pro-inflammatory and chemotactic cytokines together with their receptors associate with this disorder. Interleukin-2 receptor gamma (IL-2RG) represents an important signaling component of many interleukin receptors and so far, no data on the functional state of this receptor in schizophrenia have been reported. The aim of this study was to investigate mRNA expression of the IL2RG gene (IL2RG) in schizophrenia patients in comparison with healthy subjects (controls). Total RNA was isolated from peripheral blood of 66 schizophrenia patients and 99 healthy subjects of Armenian population. The mRNA expression was determined by quantitative real-time polymerase chain reaction (RT-PCR) using PSMB2 as housekeeping gene. IL2RG mRNA expression was upregulated in peripheral blood of patients in comparison with controls (patients vs. controls, median [interquartile range]: 2.080 [3.428-1.046] vs. 0.324 [0.856-0.000], p<0.0001). In conclusion, our findings suggest that over-expression of the IL2RG gene may be implicated in altered immune response in schizophrenia and contribute to the pathomechanisms of this disorder.