- Mycemycins A-E, New Dibenzoxazepinones Isolated from Two Different Streptomycetes.
Mycemycins A-E, New Dibenzoxazepinones Isolated from Two Different Streptomycetes.
Marine drugs (2015-10-06)
Ning Liu, Fangying Song, Fei Shang, Ying Huang
PMID26437421
ABSTRACT
Five new dibenzoxazepinone derivatives, mycemycins A-E (1-5), were isolated from the ethanol extracts of mycelia of two different streptomycetes. 1 and 2 were isolated from an acidic red soil-derived strain, Streptomyces sp. FXJ1.235, and 3-5 from a gntR gene-disrupted deep-sea strain named Streptomyces olivaceus FXJ8.012Δ1741. The structures of mycemycins were elucidated by a combination of spectroscopic analyses, including 1D- and 2D-NMR techniques.
Product Description
Roche
Reverse Transcriptase Assay, colorimetric, suitable for enzyme immunoassay, kit of 1 (14 components), sufficient for ≤200 tests
Pricing and availability is not currently available.