Merck
  • Muscle-specific mRNA isoform encodes a protein composed mainly of the N-terminal 175 residues of type 2 Ins(1,4,5)P3 receptor.
Muscle-specific mRNA isoform encodes a protein composed mainly of the N-terminal 175 residues of type 2 Ins(1,4,5)P3 receptor.
The Biochemical journal (1998-09-08)
A Futatsugi, G Kuwajima, K Mikoshiba
ABSTRACT
We have found a novel isoform of the mouse type 2 Ins(1,4,5)P3 receptor [Ins(1,4,5)P3R] mRNA by reverse transcriptase-mediated PCR analysis. The novel isoform, which was expressed specifically in skeletal muscle and heart, was generated by the inclusion of a novel exon. As this exon contains a stop codon, the isoform encodes a putative protein (designated TIPR) consisting of 175 acid residues of the type 2 Ins(1,4,5)P3R and the following six residues derived from this exon. We transfected the cDNA of this isoform into COS-7 cells; these cells expressed a 24 kDa protein that was recognized by an antibody against TIPR produced in Escherichia coli. The isoform encoding TIPR was also found in human skeletal muscle and heart. The N-terminal region of Ins(1,4,5)P3R is suggested to have a role in ligand binding and to interact with the C-terminal channel domain of Ins(1,4,5)P3R itself. TIPR might regulate the Ins(1,4,5)P3 signal pathway in both muscles.