Merck
  • p38 MAPK activation upregulates proinflammatory pathways in skeletal muscle cells from insulin-resistant type 2 diabetic patients.
p38 MAPK activation upregulates proinflammatory pathways in skeletal muscle cells from insulin-resistant type 2 diabetic patients.
American journal of physiology. Endocrinology and metabolism (2014-11-06)
Audrey E Brown, Jane Palsgaard, Rehannah Borup, Peter Avery, David A Gunn, Pierre De Meyts, Stephen J Yeaman, Mark Walker
ABSTRACT
Skeletal muscle is the key site of peripheral insulin resistance in type 2 diabetes. Insulin-stimulated glucose uptake is decreased in differentiated diabetic cultured myotubes, which is in keeping with a retained genetic/epigenetic defect of insulin action. We investigated differences in gene expression during differentiation between diabetic and control muscle cell cultures. Microarray analysis was performed using skeletal muscle cell cultures established from type 2 diabetic patients with a family history of type 2 diabetes and clinical evidence of marked insulin resistance and nondiabetic control subjects with no family history of diabetes. Genes and pathways upregulated with differentiation in the diabetic cultures, compared with controls, were identified using Gene Spring and Gene Set Enrichment Analysis. Gene sets upregulated in diabetic myotubes were associated predominantly with inflammation. p38 MAPK was identified as a key regulator of the expression of these proinflammatory gene sets, and p38 MAPK activation was found to be increased in the diabetic vs. control myotubes. Although inhibition of p38 MAPK activity decreased cytokine gene expression from the cultured diabetic myotubes significantly, it did not improve insulin-stimulated glucose uptake. Increased cytokine expression driven by increased p38 MAPK activation is a key feature of cultured myotubes derived from insulin-resistant type 2 diabetic patients. p38 MAPK inhibition decreased cytokine expression but did not affect the retained defect of impaired insulin action in the diabetic muscle cells.