Merck
  • "(+)-And (-)-borneol: efficacious positive modulators of GABA action at human recombinant a 1 ? 2 ? 2L GABA A receptors.
"(+)-And (-)-borneol: efficacious positive modulators of GABA action at human recombinant a 1 ? 2 ? 2L GABA A receptors.
Biochemical Pharmacology (2005)
Granger, Renee E., Erica L. Campbell, and Graham AR Johnston.