Merck
  • High-resolution proteomics unravel architecture and molecular diversity of native AMPA receptor complexes.
High-resolution proteomics unravel architecture and molecular diversity of native AMPA receptor complexes.
Neuron (2012-05-29)
Jochen Schwenk, Nadine Harmel, Aline Brechet, Gerd Zolles, Henrike Berkefeld, Catrin Swantje Müller, Wolfgang Bildl, David Baehrens, Björn Hüber, Akos Kulik, Nikolaj Klöcker, Uwe Schulte, Bernd Fakler
ABSTRACT
AMPA-type glutamate receptors (AMPARs) are responsible for a variety of processes in the mammalian brain including fast excitatory neurotransmission, postsynaptic plasticity, or synapse development. Here, with comprehensive and quantitative proteomic analyses, we demonstrate that native AMPARs are macromolecular complexes with a large molecular diversity. This diversity results from coassembly of the known AMPAR subunits, pore-forming GluA and three types of auxiliary proteins, with 21 additional constituents, mostly secreted proteins or transmembrane proteins of different classes. Their integration at distinct abundance and stability establishes the heteromultimeric architecture of native AMPAR complexes: a defined core with a variable periphery resulting in an apparent molecular mass between 0.6 and 1 MDa. The additional constituents change the gating properties of AMPARs and provide links to the protein dynamics fundamental for the complex role of AMPARs in formation and operation of glutamatergic synapses.
Sigma-Aldrich
Anti-GSG1L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Glutamate Receptor 4 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Stargazin/Cacng2 Antibody, CT, Upstate®, from rabbit
Sigma-Aldrich
Anti-GluR2/3 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4, clone 6C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutamate Receptor 2 & 4 Antibody, clone 3A11, clone 3A11, Chemicon®, from mouse