Merck
  • Synthesis of efficient gene delivery systems by grafting pegylated alkylcarboxylate chains to PAMAM dendrimers: Evaluation of transfection efficiency and cytotoxicity in cancerous and mesenchymal stem cells.
Synthesis of efficient gene delivery systems by grafting pegylated alkylcarboxylate chains to PAMAM dendrimers: Evaluation of transfection efficiency and cytotoxicity in cancerous and mesenchymal stem cells.
Journal of biomaterials applications (2015-08-13)
Sara Ayatollahi, Maryam Hashemi, Reza Kazemi Oskuee, Zahra Salmasi, Ahad Mokhtarzadeh, Mona Alibolandi, Khalil Abnous, Mohammad Ramezani
ABSTRACT
The applications of polyamidoamine (PAMAM) dendrimers have attracted much attention in biomedicine specially non-viral gene delivery because of thier unique characteristics including hyperbranching, multivalency, and well-defined uniform globular three-dimensional structures. In the current study, in order to enhance the transfection efficiency and reduce the cytotoxicity of PAMAMs, bromoalkylcarboxylates with different chain length (2-bromoacetic, 6-bromohexanoic, 10-bromodecanoic and 16-bromohexadecanoic acids) were covalently conjugated with 10% and 30% of primary amines of generation 4 and 5 (G4 and G5) of PAMAM dendrimers to increase the hydrophobicity of the carrier. At the next stage, the alkylcarboxylate-PAMAMs were pegylaed to further modify the PAMAM structures for biological applications. Obtained results demonstrated that the prepared PAMAM derivatives had particle size around 140 nm with net-positive surface charge. None of the prepared PAMAM-based non-viral vactors exhibited significant hemolytic activity and also cytotoxicity. Meanwhile decahexanoate-PAMAM G4 [G4(16C-10%)] and decanoate-PAMAM G4 conjugated to polyethylene glycol (PEG) (G4[(10C-30%)(10C-PEG)]) showed highest transfection efficiency in murine neuroblastoma (Neuro-2a) cell line, interestingly only the latter had improved transfection efficiency in mesenchymal stem cells (MSCs). This study proved the potential utility of alkylcarboxylate-grafted PAMAM dendrimers (G4 and G5) with or without PEG modification for efficient gene transfer into cancerous cells as well as MSCs.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
10-Bromodecanoic acid, 95%