Merck
  • Incorporation of liposomes containing squid tunic ACE-inhibitory peptides into fish gelatin.
Incorporation of liposomes containing squid tunic ACE-inhibitory peptides into fish gelatin.
Journal of the science of food and agriculture (2015-02-24)
Mauricio Mosquera, Begoña Giménez, Pilar Montero, Maria Carmen Gómez-Guillén
ABSTRACT
Hydrolysates from collagen of jumbo squid (Dosidicus gigas) tunics have shown excellent angiotensin I-converting enzyme (ACE)-inhibitory activity. However, peptides directly included in food systems may suffer a decrease in activity, which could be minimized by loading them into nanoliposomes. A fraction of peptides with molecular weights <1 kDa obtained from hydrolyzed squid tunics, with reasonably high ACE-inhibitory activity (half-maximal inhibitory concentration IC50 = 0.096 g L(-1)), was encapsulated in phosphatidylcholine nanoliposomes. The peptide concentration affected the encapsulation efficiency and the stability of the resulting liposomes, which remained with a high zeta potential value (-54.3 mV) for at least 1 week at the most suitable peptide concentration. The optimal peptide concentration was established as 1.75 g L(-1). Liposomes obtained with this peptide concentration showed an encapsulation efficiency of 53%, a zeta potential of -59 mV, an average diameter of 70.3 nm and proved to be stable in the pH range 3-7 at 4 °C. Liposomes containing ACE-inhibitory peptides were incorporated in fish gelatin without detriment to the rheological properties and thermal stability of the resulting cold-induced gel. The ACE-inhibitory activity of the peptide fraction, which was not affected by the encapsulation process, conferred the bioactive potential to the nanoliposome-containing gelatin gel.
Sigma-Aldrich
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)