Merck
  • Intramolecular chalcogen-tin interactions in [(o-MeEC6H4)CH2]2SnPh(2-n)Cl(n) (E = S, O, CH2; n = 0, 1, 2) and intermolecular chlorine-tin interactions in the meta- and para-methoxy isomers.
Intramolecular chalcogen-tin interactions in [(o-MeEC6H4)CH2]2SnPh(2-n)Cl(n) (E = S, O, CH2; n = 0, 1, 2) and intermolecular chlorine-tin interactions in the meta- and para-methoxy isomers.
Inorganic chemistry (2010-01-06)
Diana Gabriela Vargas-Pineda, Tanya Guardado, Francisco Cervantes-Lee, Alejandro J Metta-Magana, Keith H Pannell
ABSTRACT
Organotin(IV) compounds of the type [(o-MeEC(6)H(4))CH(2)](2)SnPh(2-n)Cl(n) were synthesized, E = O, n = 0 (1), n = 1 (2), and n = 2 (3); E = S, n = 0 (4), n = 1 (5), and n = 2 (6); and E = CH(2), n = 0 (7), n = 1 (8), and n = 2 (9). The dichloro compounds 3 and 6 have been investigated by single-crystal X-ray diffraction and exhibit bicapped tetrahedral geometry at the tin atom as a consequence of significant intramolecular Sn...O (3) and Sn...S (6) secondary bonding, in monomolecular units. Compound 3, when crystallized from a hexane/THF solvent mixture, shows two different conformers, 3' and 3'', in the crystal structure; 3' has two equivalent Sn...O interactions, while 3'' has two nonequivalent Sn...O interactions. Upon the recrystallization of 3 from hexane, only a single structural form is observed, 3'. The Sn...E distances in 3', 3'', and 6 are 71.3, 73.5 and 72.9, and 76.3% of the SigmavdW radii, respectively. The meta- and para-substituted isomers of 3 (10, 11) exhibit a distortion at the tin atom due to self-association via intermolecular Sn...Cl interactions, resulting in polymeric structures. (119)Sn NMR spectroscopy suggests that the intramolecular Sn...E interactions persist in solution for the dichloride compounds 3 and 6.