Merck
  • A new class of nonhormonal pregnancy-terminating agents. Synthesis and contragestational activity of 3,5-diaryl-s-triazoles.
A new class of nonhormonal pregnancy-terminating agents. Synthesis and contragestational activity of 3,5-diaryl-s-triazoles.
Journal of medicinal chemistry (1983-08-01)
A Omodei-Salé, P Consonni, G Galliani
ABSTRACT
A series of 3,5-diaryl-s-triazoles were synthesized and evaluated as postimplantation contragestational agents. The introduction of various substituents (e.g., an o-alkyl chain on one phenyl and a m-alkoxy group on the other) was found to increase the potency. Several compounds with very high pregnancy-terminating activity in both hamsters and rats were obtained. One of these, 3-(2-ethylphenyl)-5-(3-methoxyphenyl)-s-triazole, DL 111 (36), was selected for detailed evaluation in various animal species. A synthetic scheme for the preparation of these compounds and preliminary structure-activity relationships are presented.