Merck
  • A novel mechanism of keratin cytoskeleton organization through casein kinase Iα and FAM83H in colorectal cancer.
A novel mechanism of keratin cytoskeleton organization through casein kinase Iα and FAM83H in colorectal cancer.
Journal of cell science (2013-08-02)
Takahisa Kuga, Hideaki Kume, Naoko Kawasaki, Misako Sato, Jun Adachi, Takashi Shiromizu, Isamu Hoshino, Takanori Nishimori, Hisahiro Matsubara, Takeshi Tomonaga
ABSTRACT
Keratin filaments form cytoskeletal networks in epithelial cells. Dynamic rearrangement of keratin filament networks is required for epithelial cells to perform cellular processes such as cell migration and polarization; however, the mechanism governing keratin filament rearrangement remains unclear. Here, we describe a novel mechanism of keratin cytoskeleton organization mediated by casein kinase Iα (CK-1α) and a newly identified keratin-associated protein, FAM83H. Knockdown of FAM83H induces keratin filament bundling, whereas overexpression of FAM83H disassembles keratin filaments, suggesting that FAM83H regulates the filamentous state of keratins. Intriguingly, keratin filament bundling is concomitant with the dissociation of CK-1α from keratin filaments, whereas aberrant speckle-like localization of CK-1α is observed concomitantly with keratin filament disassembly. Furthermore, CK-1α inhibition, similar to FAM83H knockdown, causes keratin filament bundling and reverses keratin filament disassembly induced by FAM83H overexpression, suggesting that CK-1α mediates FAM83H-dependent reorganization of keratin filaments. Because the N-terminal region of FAM83H interacts with CK-1α and the C-terminal region interacts with keratins, FAM83H might tether CK-1α to keratins. Colorectal cancer tissue also shows keratin filament disassembly accompanied with FAM83H overexpression and aberrant CK-1α localization, and FAM83H-overexpressing cancer cells exhibit loss or alteration of epithelial cell polarity. Importantly, knockdown of FAM83H inhibits cell migration accompanied by keratin cytoskeleton rearrangement in colorectal cancer cells. These results suggest that keratin cytoskeleton organization is regulated by FAM83H-mediated recruitment of CK-1α to keratins, and that keratin filament disassembly caused by overexpression of FAM83H and aberrant localization of CK-1α could contribute to the progression of colorectal cancer.
Sigma-Aldrich
Anti-FAM83H antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Pricing and availability is not currently available.