Merck
Acylcarnitine analogues as topical, microbicidal spermicides.
Bioorganic & medicinal chemistry letters (1999-09-25)
P S Savle, G F Doncel, S D Bryant, M P Hubieki, R G Robinette, R D Gandour
ABSTRACT
Acylcarnitine analogues, (+)-6-Carboxylatomethyl-2-alkyl-4,4-dimethylmorpholinium (Z-n, where n = the number of carbons in the alkyl chain), synthesized in multi-gram quantities show in vitro activities as spermicides, anti-HIV agents, and inhibitors of the growth of Candida albicans. Activity improves with increasing chain length. Compound Z-15 is a candidate for further study as a topical, microbicidal spermicide.