Merck
  • Immobilized alpha-diazophosphonoacetate as a versatile key precursor for palladium catalyzed indole synthesis on a polymer support.
Immobilized alpha-diazophosphonoacetate as a versatile key precursor for palladium catalyzed indole synthesis on a polymer support.
Chemical communications (Cambridge, England) (2002-07-18)
Kazuo Yamazaki, Yoshinori Kondo
ABSTRACT
Rh(II)-catalyzed N-H insertion reaction of immobilized alpha-diazophosphonoacetate with 2-haloanilines followed by Horner-Emmons reaction gave immobilized enaminoesters, which were efficiently cyclized to indoles via intramolecular palladium catalyzed reaction on a polymer support.