Merck
  • Simultaneous separation and sensitive determination of free fatty acids in blood plasma by high-performance liquid chromatography.
Simultaneous separation and sensitive determination of free fatty acids in blood plasma by high-performance liquid chromatography.
Journal of chromatography (1985-12-13)
I Yanagisawa, M Yamane, T Urayama
ABSTRACT
The quantitative determination of saturated and unsaturated fatty acids (ranging from acetic acid to lignoceric acid) in biological samples is presented. The secondary amine group of 5-(dimethylamino)-1-naphthalenesulponyl-semipiperazide (dansyl-semipiperazide) reacts with the carboxyl group of the fatty acids to form an amide linkage in order to obtain fluorescent derivatives of the acids. The fluorescent derivatives are analysed by high-performance liquid chromatography (HPLC) using an internal standard.