Merck
  • Serum hydroxyl radical scavenging capacity as quantified with iron-free hydroxyl radical source.
Serum hydroxyl radical scavenging capacity as quantified with iron-free hydroxyl radical source.
Journal of clinical biochemistry and nutrition (2009-10-02)
Nobuyuki Endo, Shigeru Oowada, Yoshimi Sueishi, Masashi Shimmei, Keisuke Makino, Hirotada Fujii, Yashige Kotake
ABSTRACT
We have developed a simple ESR spin trapping based method for hydroxyl (OH) radical scavenging-capacity determination, using iron-free OH radical source. Instead of the widely used Fenton reaction, a short (typically 5 seconds) in situ UV-photolysis of a dilute hydrogen peroxide aqueous solution was employed to generate reproducible amounts of OH radicals. ESR spin trapping was applied to quantify OH radicals; the decrease in the OH radical level due to the specimen's scavenging activity was converted into the OH radical scavenging capacity (rate). The validity of the method was confirmed in pure antioxidants, and the agreement with the previous data was satisfactory. In the second half of this work, the new method was applied to the sera of chronic renal failure (CRF) patients. We show for the first time that after hemodialysis, OH radical scavenging capacity of the CRF serum was restored to the level of healthy control. This method is simple and rapid, and the low concentration hydrogen peroxide is the only chemical added to the system, that could eliminate the complexity of iron-involved Fenton reactions or the use of the pulse-radiolysis system.