Merck
  • PEG-supported synthesis of pyrazole oligoamides with peptide beta-sheet affinity.
PEG-supported synthesis of pyrazole oligoamides with peptide beta-sheet affinity.
Organic & biomolecular chemistry (2004-05-27)
Katerina Cernovská, Miriam Kemter, Hans-Christoph Gallmeier, Petra Rzepecki, Thomas Schrader, Burkhard König
ABSTRACT
Pyrazole amino acid oligoamides were prepared on polyethylene glycol starting from nitro pyrazole carboxylic acids or protected pyrazole amino acids. The polymer support facilitates product isolation during synthesis and makes the target oligoamides soluble in chloroform and water. This allows the determination of their binding properties towards peptides. Moderate affinity, which increases with the number of pyrazole units, is observed in chloroform and water.