Merck
  • Comparison of myxobacterial diversity and evaluation of isolation success in two niches: Kiritimati Island and German compost.
Comparison of myxobacterial diversity and evaluation of isolation success in two niches: Kiritimati Island and German compost.
MicrobiologyOpen (2015-12-17)
Kathrin I Mohr, Marc Stechling, Joachim Wink, Elke Wilharm, Marc Stadler
ABSTRACT
Myxobacteria harbor an enormous potential for new bioactive secondary metabolites and therefore the isolation of in particular new groups is of great interest. The diversity of myxobacteria present in two ecological habitats, namely sand from Kiritimati Island and German compost, was evaluated by both cultivation-based and cultivation-independent methods. Phylogenetic analyses of the strains in comparison with 16S rRNA gene sequences from cultured and uncultured material in GenBank revealed a great potential of undescribed myxobacteria in both sampling sites. Several OTUs (operational taxonomic units) represent unknown taxa and were detected by clone bank analyses, but not by cultivation. Clone bank analyses indicated that the myxobacterial community is predominantly indigenous. The 16S rDNA libraries from the two samples were generated from total community DNA with myxobacterial specific forward and universal reverse primer sets. The clones were partially sequenced. Cultivation was successful for exclusively bacteriolytic, but not for cellulolytic myxobacteria and revealed 42 strains from the genera Corallococcus, Myxococcus, and Polyangium. The genera of Myxococcaceae family were represented by both approaches. But, even in this well studied family, as well as in the suborders Sorangiineae and Nannocystineae, a considerable number of clones were assigned to, if any, uncultivated organisms. Our study shows an overrepresentation of the genera Myxococcus spp. and Corallococcus spp. with standard cultivation methods. However, high deficits are demonstrated in the cultivation success of the myxobacterial diversity detected by exclusively cultivation-independent approaches. Especially, clades which are exclusively represented by clones are of high interest with regard to the cultivation of new bioactive secondary metabolite producers.
Sigma-Aldrich
Texan reagent reservoir for multichannel pipettes, with lid, non-sterile, pack of 100 ea (10 pavks of 10ea)
Pricing and availability is not currently available.