Merck
  • Antagonistic effects of beta-site amyloid precursor protein-cleaving enzymes 1 and 2 on beta-amyloid peptide production in cells.
Antagonistic effects of beta-site amyloid precursor protein-cleaving enzymes 1 and 2 on beta-amyloid peptide production in cells.
The Journal of biological chemistry (2003-06-13)
Guriqbal Basi, Normand Frigon, Robin Barbour, Tam Doan, Grace Gordon, Lisa McConlogue, Sukanto Sinha, Michelle Zeller
ABSTRACT
The deposition of extracellular beta-amyloid peptide (A beta) in the brain is a pathologic feature of Alzheimer's disease. The beta-site amyloid precursor protein cleaving enzyme 1 (BACE1), an integral membrane aspartyl protease responsible for cleavage of amyloid precursor protein (APP) at the beta-site, promotes A beta production. A second integral membrane aspartyl protease related to BACE1, referred to as beta-site amyloid precursor protein cleaving enzyme 2 (BACE2) has also been demonstrated to cleave APP at the beta-cleavage site in transfected cells. The role of endogenous BACE2 in A beta production remains unresolved. We investigated the role of endogenous BACE2 in A beta production in cells by selective inactivation of its transcripts using RNA interference. We are able to reduce steady state levels for mRNA for each enzyme by >85%, and protein amounts by 88-94% in cells. Selective inactivation of BACE1 by RNA interference results in decreased beta-cleaved secreted APP and A beta peptide secretion from cells, as expected. Selective inactivation of BACE2 by RNAi results in increased beta-cleaved secreted APP and A beta peptide secretion from cells. Simultaneous targeting of both enzymes by RNA interference does not have any net effect on A beta released from cells. Our observations of changes in APP metabolism and A beta are consistent with a role of BACE2 in suppressing A beta production in cells that co-express both enzymes.