Merck
UNC2025, a potent and orally bioavailable MER/FLT3 dual inhibitor.
Journal of medicinal chemistry (2014-07-30)
Weihe Zhang, Deborah DeRyckere, Debra Hunter, Jing Liu, Michael A Stashko, Katherine A Minson, Christopher T Cummings, Minjung Lee, Trevor G Glaros, Dianne L Newton, Susan Sather, Dehui Zhang, Dmitri Kireev, William P Janzen, H Shelton Earp, Douglas K Graham, Stephen V Frye, Xiaodong Wang
ABSTRACT
We previously reported a potent small molecule Mer tyrosine kinase inhibitor UNC1062. However, its poor PK properties prevented further assessment in vivo. We report here the sequential modification of UNC1062 to address DMPK properties and yield a new potent and highly orally bioavailable Mer inhibitor, 11, capable of inhibiting Mer phosphorylation in vivo, following oral dosing as demonstrated by pharmaco-dynamic (PD) studies examining phospho-Mer in leukemic blasts from mouse bone marrow. Kinome profiling versus more than 300 kinases in vitro and cellular selectivity assessments demonstrate that 11 has similar subnanomolar activity against Flt3, an additional important target in acute myelogenous leukemia (AML), with pharmacologically useful selectivity versus other kinases examined.
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)