Merck
  • Photoinitiated polymerization of 2-hydroxyethyl methacrylate by riboflavin/triethanolamine in aqueous solution: a kinetic study.
Photoinitiated polymerization of 2-hydroxyethyl methacrylate by riboflavin/triethanolamine in aqueous solution: a kinetic study.
ISRN pharmaceutics (2013-11-01)
Iqbal Ahmad, Kefi Iqbal, Muhammad Ali Sheraz, Sofia Ahmed, Tania Mirza, Sadia Hafeez Kazi, Mohammad Aminuddin
ABSTRACT
The polymerization of 1-3 M 2-hydroxyethyl methacrylate (HEMA) initiated by riboflavin/triethanolamine system has been studied in the pH range 6.0-9.0. An approximate measure of the kinetics of the reaction during the initial stages (~5% HEMA conversion) has been made to avoid the effect of any variations in the volume of the medium. The concentration of HEMA in polymerized solutions has been determined by a UV spectrophotometric method at 208 nm with a precision of ±3%. The initial rate of polymerization of HEMA follows apparent first-order kinetics and the rates increase with pH. This may be due to the presence of a labile proton on the hydroxyl group of HEMA. The second-order rate constants for the interaction of triethanolamine and HEMA lie in the range of 2.36 to 8.67 × 10(-2) M(-1) s(-1) at pH 6.0-9.0 suggesting an increased activity with pH. An increase in the viscosity of HEMA solutions from 1 M to 3 M leads to a decrease in the rate of polymerization probably as a result of the decrease in the reactivity of the flavin triplet state. The effect of pH and viscosity of the medium on the rate of reaction has been evaluated.
Sigma-Aldrich
Lumichrome
Pricing and availability is not currently available.