Merck
  • Endoplasmic reticulum stress signaling transmitted by ATF6 mediates apoptosis during muscle development.
Endoplasmic reticulum stress signaling transmitted by ATF6 mediates apoptosis during muscle development.
The Journal of cell biology (2005-05-18)
Keiko Nakanishi, Tatsuhiko Sudo, Nobuhiro Morishima
ABSTRACT
Although apoptosis occurs during myogenesis, its mechanism of initiation remains unknown. In a culture model, we demonstrate activation of caspase-12, the initiator of the endoplasmic reticulum (ER) stress-specific caspase cascade, during apoptosis associated with myoblast differentiation. Induction of ER stress-responsive proteins (BiP and CHOP) was also observed in both apoptotic and differentiating cells. ATF6, but not other ER stress sensors, was specifically activated during apoptosis in myoblasts, suggesting that partial but selective activation of ER stress signaling was sufficient for induction of apoptosis. Activation of caspase-12 was also detected in developing muscle of mouse embryos and gradually disappeared later. CHOP was also transiently induced. These results suggest that specific ER stress signaling transmitted by ATF6 leads to naturally occurring apoptosis during muscle development.