Merck
  • Targeted lipidomics in Drosophila melanogaster identifies novel 2-monoacylglycerols and N-acyl amides.
Targeted lipidomics in Drosophila melanogaster identifies novel 2-monoacylglycerols and N-acyl amides.
PloS one (2013-07-23)
Giuseppe Tortoriello, Brandon P Rhodes, Sara M Takacs, Jordyn M Stuart, Arjun Basnet, Siham Raboune, Theodore S Widlanski, Patrick Doherty, Tibor Harkany, Heather B Bradshaw
ABSTRACT
Lipid metabolism is critical to coordinate organ development and physiology in response to tissue-autonomous signals and environmental cues. Changes to the availability and signaling of lipid mediators can limit competitiveness, adaptation to environmental stressors, and augment pathological processes. Two classes of lipids, the N-acyl amides and the 2-acyl glycerols, have emerged as important signaling molecules in a wide range of species with important signaling properties, though most of what is known about their cellular functions is from mammalian models. Therefore, expanding available knowledge on the repertoire of these lipids in invertebrates will provide additional avenues of research aimed at elucidating biosynthetic, metabolic, and signaling properties of these molecules. Drosophila melanogaster is a commonly used organism to study intercellular communication, including the functions of bioactive lipids. However, limited information is available on the molecular identity of lipids with putative biological activities in Drosophila. Here, we used a targeted lipidomics approach to identify putative signaling lipids in third instar Drosophila larvae, possessing particularly large lipid mass in their fat body. We identified 2-linoleoyl glycerol, 2-oleoyl glycerol, and 45 N-acyl amides in larval tissues, and validated our findings by the comparative analysis of Oregon-RS, Canton-S and w1118 strains. Data here suggest that Drosophila represent another model system to use for the study of 2-acyl glycerol and N-acyl amide signaling.