Merck
  • Cell surface attachment structures contribute to biofilm formation and xylem colonization by Erwinia amylovora.
Cell surface attachment structures contribute to biofilm formation and xylem colonization by Erwinia amylovora.
Applied and environmental microbiology (2011-08-09)
Jessica M Koczan, Bryan R Lenneman, Molly J McGrath, George W Sundin
ABSTRACT
Biofilm formation plays a critical role in the pathogenesis of Erwinia amylovora and the systemic invasion of plant hosts. The functional role of the exopolysaccharides amylovoran and levan in pathogenesis and biofilm formation has been evaluated. However, the role of biofilm formation, independent of exopolysaccharide production, in pathogenesis and movement within plants has not been studied previously. Evaluation of the role of attachment in E. amylovora biofilm formation and virulence was examined through the analysis of deletion mutants lacking genes encoding structures postulated to function in attachment to surfaces or in cellular aggregation. The genes and gene clusters studied were selected based on in silico analyses. Microscopic analyses and quantitative assays demonstrated that attachment structures such as fimbriae and pili are involved in the attachment of E. amylovora to surfaces and are necessary for the production of mature biofilms. A time course assay indicated that type I fimbriae function earlier in attachment, while type IV pilus structures appear to function later in attachment. Our results indicate that multiple attachment structures are needed for mature biofilm formation and full virulence and that biofilm formation facilitates entry and is necessary for the buildup of large populations of E. amylovora cells in xylem tissue.
Product Description
Roche
Expand Long Range dNTPack, suitable for PCR, hotstart: no, Long Range PCR
Pricing and availability is not currently available.