Merck
  • Detection of bphAa gene expression of Rhodococcus sp. strain RHA1 in soil using a new method of RNA preparation from soil.
Detection of bphAa gene expression of Rhodococcus sp. strain RHA1 in soil using a new method of RNA preparation from soil.
Bioscience, biotechnology, and biochemistry (2008-03-08)
Yong Wang, Jun Shimodaira, Tomomichi Miyasaka, Sho Morimoto, Takanori Oomori, Naoto Ogawa, Masao Fukuda, Takeshi Fujii
ABSTRACT
To understand the response of soil bacteria to the surrounding environment, it is necessary to examine the gene expression profiles of the bacteria in the soil. For this purpose, we developed a new method of extracting RNA from soil reproducibly. Using this new method, we extracted RNA from a field soil, which was sterilized and inoculated with Rhodococcus sp. strain RHA1, a biphenyl degrader isolated from gamma-hexachlorocyclohexane-contaminated soil. Data from agarose gel electrophoresis indicated that the extracted RNA was purified properly. This new method can be applied easily in the preparation of large amounts of RNA. Real-time reverse transcription-polymerase chain reaction (RT-PCR) experiments performed by the TaqMan method suggested that the bphAa gene in this strain, which is involved in the degradation of biphenyl, was induced in the biphenyl amended soil.