Merck
  • Mechanisms of resistance to trimethoprim, the sulfonamides, and trimethoprim-sulfamethoxazole.
Mechanisms of resistance to trimethoprim, the sulfonamides, and trimethoprim-sulfamethoxazole.
Reviews of infectious diseases (1982-03-01)
R L Then
ABSTRACT
A variety of different mechanisms are known to be responsible for either natural or acquired resistance to trimethoprim, the sulfonamides, or trimethoprim-sulfonamide combinations. Some mechanisms of obvious clinical importance have been studied intensively. Among these are unique bypass mechanisms such as the synthesis of drug-resistant, plasmid-coded dihydrofolate reductase or dihydropteroate synthetase; such mechanisms so far have not been encountered in studies of resistance to other drugs. This article focuses on several mechanisms of resistance that have rarely been discussed in the past, including metabolic alteration of trimethoprim or the sulfonamides and hyperproduction of p-aminobenzoic acid, and on the simultaneous presence of more than one mechanism. The role of these mechanisms in the resistance of clinical isolates requires further investigation.
Product Description
USP
Sulfamethoxazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Sulfamethoxazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sulfamethoxazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Sulfamethoxazole
Supelco
Sulfathiazole
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Supelco
Sulfamethoxazole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Supelco
Sulfadimethoxine, 98.0-102.0%