Merck
  • Paclitaxel from primary taxanes: a perspective on creative invention in organozirconium chemistry.
Paclitaxel from primary taxanes: a perspective on creative invention in organozirconium chemistry.
The Journal of organic chemistry (2007-03-27)
Bruce Ganem, Roland R Franke
ABSTRACT
In this Perspective, which describes the achievements recognized by the 2007 ACS Award for Creative Invention, we discuss the discovery of a new synthetic reaction and its translation into a substantially improved method for manufacturing a major pharmaceutical product--the blockbuster anticancer drug, paclitaxel. The role of creativity in the discovery and invention processes is also discussed. As is often the case, chance discovery and serendipitous findings played a role in the evolution of this work. Translation of the basic research into a commercially viable paclitaxel semisynthesis is also described. The final manufacturing process illustrates the enormous impact that the globalization of markets has had on chemical and pharmaceutical manufacturing.
Sigma-Aldrich
Bis(cyclopentadienyl)zirconium(IV) chloride hydride, 95%
Pricing and availability is not currently available.