Merck
  • Isolation and identification of an antifungal naphthopyran derivative from Rhinacanthus nasutus.
Isolation and identification of an antifungal naphthopyran derivative from Rhinacanthus nasutus.
Journal of natural products (1993-02-01)
O Kodama, H Ichikawa, T Akatsuka, V Santisopasri, A Kato, Y Hayashi
ABSTRACT
Rhinacanthus nasutus (Acanthaceae) gave a new antifungal naphthopyran derivative. The structure of the compound was elucidated by spectroscopic methods as 3,4-dihydro-3,3-dimethyl-2H-naphtho[2,3-b]pyran-5,10-dione [1].