Merck
  • Simple, stable, and versatile double-allylation reagents for the stereoselective preparation of skeletally diverse compounds.
Simple, stable, and versatile double-allylation reagents for the stereoselective preparation of skeletally diverse compounds.
Journal of the American Chemical Society (2007-02-24)
Feng Peng, Dennis G Hall
Sigma-Aldrich
(1R,2R,3S,5R)-(−)-Pinanediol, 99%