Merck
  • Overexpressed CacyBP/SIP leads to the suppression of growth in renal cell carcinoma.
Overexpressed CacyBP/SIP leads to the suppression of growth in renal cell carcinoma.
Biochemical and biophysical research communications (2007-04-03)
Shiren Sun, Xiaoxuan Ning, Jie Liu, Lili Liu, Yu Chen, Shuang Han, Yanqi Zhang, Jie Liang, Kaichun Wu, Daiming Fan
ABSTRACT
Calcyclin-binding protein/Siah-1-interacting protein (CacyBP/SIP), a target protein of S100, has been identified as a component of a novel ubiquitinylation complex leading to beta-catenin degradation, which was found to be related to the malignant phenotypes of gastric cancer. However, the roles of CacyBP/SIP in renal cell carcinoma still remain unclear. In the present study, we had analyzed the expression of the CacyBP/SIP protein in human renal cancer cells and clinical tissue samples. The possible roles of CacyBP/SIP in regulating the malignant phenotype of renal cancer cells were also investigated. The results demonstrated that the expression of CacyBP/SIP was markedly down-regulated in renal cell carcinoma tissues and cell lines. Ectopic overexpression of CacyBP/SIP in A498 cells inhibited the proliferation of this cell and delayed cell cycle progression significantly, which might be related to the down-regulation of Cyclin D1 through reducing beta-catenin protein. CacyBP/SIP also suppressed colony formation in soft agar and its tumorigenicity in nude mice. Taken together, our work showed that CacyBP/SIP, as a novel down-regulated gene in renal cell carcinoma, suppressed proliferation and tumorigenesis of renal cancer cells.