Merck
Developing imidazoles as CEST MRI pH sensors.
Contrast media & molecular imaging (2016-04-14)
Xing Yang, Xiaolei Song, Sangeeta Ray Banerjee, Yuguo Li, Youngjoo Byun, Guanshu Liu, Zaver M Bhujwalla, Martin G Pomper, Michael T McMahon
ABSTRACT
A series of intra-molecular hydrogen bonded imidazoles and related heterocyclic compounds were screened for their N-H chemical exchange saturation transfer (CEST) magnetic resonance imaging (MRI) contrast properties. Of the compounds, imidazole-4,5-dicarboxamides (I45DCs) were found to provide the strongest contrast, with the contrast produced at a large chemical shift from water (7.8 ppm) and strongly dependent on pH. We have tested several probes based on this scaffold, and demonstrated that these probes could be applied for in vivo detection of kidney pH after intravenous administration. Copyright © 2016 John Wiley & Sons, Ltd.