Merck
  • Vasorelaxant Action of the Chloroform Fraction of Orthosiphon stamineus via NO/cGMP Pathway, Potassium and Calcium Channels.
Vasorelaxant Action of the Chloroform Fraction of Orthosiphon stamineus via NO/cGMP Pathway, Potassium and Calcium Channels.
The American journal of Chinese medicine (2016-11-05)
Mun Fei Yam, Chu Shan Tan, Mariam Ahmad, Shibao Ruan
ABSTRACT
Orthosiphon stamineus Benth. (Lamiaceae) is an important plant in traditional folk medicine that is used to treat hypertension and kidney stones. In humans, this plant has been tested as an addition regiment for antihypertensive treatment. Among the treatments for hypertension, O. stamineus had been to have diuretic and vasorelaxant effects in animal models. There is still very little information regarding the vasorelaxant effect of O. stamineus. Therefore, the present study was designed to investigate the vasorelaxant activity and mechanism of action of the fractions of O. stamineus. The vasorelaxant activity and the underlying mechanisms of the chloroform fraction of the 50% methanolic extract of O. stamineus (CF) was evaluated on thoracic aortic rings isolated from Sprague Dawley rats. CF caused relaxation of the aortic ring pre-contracted with phenylephrine in the presence and absence of endothelium, and pre-contracted with potassium chloride in endothelium-intact aortic ring. In the presence of endothelium, both indomethacin (a nonselective cyclooxygenase inhibitor) and [Formula: see text]-[1,2,4]Oxadiazolo[4,3-[Formula: see text]]quinoxalin-1-one (ODQ, selective soluble guanylate cyclase inhibitor) had a small effect on the vasorelaxation response. On the other hand, in the presence of Nω-nitro-L-arginine methyl ester (L-NAME, nitric oxide synthase inhibitor), methylene blue (cyclic guanosine monophosphate lowering agent), tetraethylammonium ([Formula: see text], nonselective calcium activator [Formula: see text] channel blocker), 4-aminopyridine (4-AP, voltage-dependent [Formula: see text] channel blocker), barium chloride ([Formula: see text], inwardly rectifying [Formula: see text] channel blocker), glibenclamide (nonspecific ATP-sensitive [Formula: see text] channel blocker), atropine (muscarinic receptor blocker) and propranolol (β-adrenergic receptor blocker), the vasorelaxant effect significantly reduced the relaxation stimulated by CF. CF was also found to be active in reducing [Formula: see text] release from the sarcoplasmic reticulum and blocking calcium channels.
Sigma-Aldrich
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder