Merck
  • Programmed necrosis - a new mechanism of steroidogenic luteal cell death and elimination during luteolysis in cows.
Programmed necrosis - a new mechanism of steroidogenic luteal cell death and elimination during luteolysis in cows.
Scientific reports (2016-12-03)
Takuo Hojo, Marta J Siemieniuch, Karolina Lukasik, Katarzyna K Piotrowska-Tomala, Agnieszka W Jonczyk, Kiyoshi Okuda, Dariusz J Skarzynski
ABSTRACT
Programmed necrosis (necroptosis) is an alternative form of programmed cell death that is regulated by receptor-interacting protein kinase (RIPK) 1 and 3-dependent, but is a caspase (CASP)-independent pathway. In the present study, to determine if necroptosis participates in bovine structural luteolysis, we investigated RIPK1 and RIPK3 expression throughout the estrous cycle, during prostaglandin F2α (PGF)-induced luteolysis in the bovine corpus luteum (CL), and in cultured luteal steroidogenic cells (LSCs) after treatment with selected luteolytic factors. In addition, effects of a RIPK1 inhibitor (necrostatin-1, Nec-1; 50 μM) on cell viability, progesterone secretion, apoptosis related factors and RIPKs expression, were evaluated. Expression of RIPK1 and RIPK3 increased in the CL tissue during both spontaneous and PGF-induced luteolysis (P < 0.05). In cultured LSCs, tumor necrosis factor α (TNF; 2.3 nM) in combination with interferon γ (IFNG; 2.5 nM) up-regulated RIPK1 mRNA and protein expression (P < 0.05). TNF + IFNG also up-regulated RIPK3 mRNA expression (P < 0.05), but not RIPK3 protein. Although Nec-1 prevented TNF + IFNG-induced cell death (P < 0.05), it did not affect CASP3 and CASP8 expression. Nec-1 decreased both RIPK1 and RIPK3 protein expression (P < 0.05). These findings suggest that RIPKs-dependent necroptosis is a potent mechanism responsible for bovine structural luteolysis induced by pro-inflammatory cytokines.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution