- Rapid ecotoxicological bioassay using delayed fluorescence in the marine cyanobacterium Cyanobium sp. (NIES-981).
Rapid ecotoxicological bioassay using delayed fluorescence in the marine cyanobacterium Cyanobium sp. (NIES-981).
Ecotoxicology (London, England) (2016-09-17)
Takahiro Yamagishi, Masakazu Katsumata, Haruyo Yamaguchi, Yohei Shimura, Masanobu Kawachi, Hiroshi Koshikawa, Yoshifumi Horie, Norihisa Tatarazako
PMID27633095
ABSTRACT
The use of delayed fluorescence intensity as an endpoint for rapid estimation of the effective concentration (EC