Merck
  • Distribution and sources of sterol biomarkers in sediments collected from a tropical estuary in Northeast Brazil.
Distribution and sources of sterol biomarkers in sediments collected from a tropical estuary in Northeast Brazil.
Environmental science and pollution research international (2016-10-04)
Morgana Frena, Ana Paula Stein Santos, Ewerton Santos, Rosianne P Silva, Michel R R Souza, Luiz A S Madureira, Marcelo R Alexandre
ABSTRACT
The Piauí-Real estuary is located along the southern coast of Sergipe state, Northeast Brazil. This estuary has great economic importance due to its physical, biological, and socioeconomic diversity, but it is subject to anthropogenic stress since the resident population in the town bordered by the estuarine system has grown in recent years. Thus, the possibility of sewage contamination originating from the approximately 450,000 inhabitants living within its drainage basin was investigated in this study. Sediment samples were collected from 15 sampling stations along the estuarine system and extracted, followed by gas chromatography-mass spectrometry (GC-MS) analysis. Six sterols were quantified, indicating natural and anthropogenic sources. Coprostanol concentrations higher than 100 ng g